Undecanoic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Hendecanoic acid
, Undecylic acid
M-Clarity™ quality level = MQ200
8.18568.1000
8.18568.0101
8.18568.1000
8.18568.0101
EM8.18568.1000CS
1160.65
USD
EM8.18568.1000
EM8.18568.0101
CA8.18568.1000
CA8.18568.0101
Undecanoic acid for synthesis, Sigma-Aldrich®
Undecanoic acid
Formula:
C₁₁H₂₂O₂ MW: 186.29 g/mol |
MDL Number:
MFCD00002730 CAS Number: 112-37-8 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 26 - 29 °C |
Molar Mass | 186.29 g/mol |
Boiling Point | 284 °C (1013 hPa) |
Flash Point |
|
Explosion Limit | 0.6 %(V) |
Density | 0.89 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...