4-(Benzyloxy)benzaldehyde for synthesis

Supplier: MilliporeSigma

8.18541.0025 8.18541.0025
EM8.18541.0025CS 8951.6 USD InStock
EM8.18541.0025 CA8.18541.0025
4-(Benzyloxy)benzaldehyde for synthesis
4-(Benzyloxy)benzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212,25 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 70 to 72 °C
Storage Temperature: ambient
MDL Number: MFCD00003387
CAS Number: 4397-53-9
EINECS: 224-527-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 70 - 72 °C
Molar Mass 212.24 g/mol
Boiling Point 197 - 199 °C (15 hPa)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR