Methyl-4-formylbenzoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Methyl 4-formylbenzoateMethyl terephthalaldehydate

M-Clarity™ quality level = MQ200

8.18490.0100 8.18490.0100
EM8.18490.0100CS 2061 USD
EM8.18490.0100 CA8.18490.0100
Methyl-4-formylbenzoate for synthesis, Sigma-Aldrich®
Methyl 4-formylbenzoate
Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00006950
CAS Number: 1571-08-0

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 56 - 59 °C
Molar Mass 164.16 g/mol
Bulk Density 528 kg/m3
Boiling Point 264 - 266 °C (1013 hPa)
Flash Point 128 °C
Density 1.2 g/cm3 (20 °C)
Ignition Point 240 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR