β-Nitrostyrene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: (1-Nitro-2-phenyl)ethylene(2-Nitrovinyl)benzeneω-Nitrostyrene

M-Clarity™ quality level = MQ200

8.18165.0025 8.18165.0025
EM8.18165.0025CS 19428.37 USD
EM8.18165.0025 CA8.18165.0025
β-Nitrostyrene for synthesis, Sigma-Aldrich®
β-Nitrostyrene
Formula: C₈H₇NO₂
MW: 149.14 g/mol
CAS Number: 102-96-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 54 - 57 °C
Molar Mass 149.14 g/mol
Boiling Point 250 - 260 °C
Ignition Point 250 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR