Allyl hexanoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hexanoic acid allyl ester

M-Clarity™ quality level = MQ200

8.14990.0250 8.14990.0250
EM8.14990.0250CS 1669 USD
CA8.14990.0250 EM8.14990.0250
Allyl hexanoate for synthesis, Sigma-Aldrich®
Allyl hexanoate
Formula: C₉H₁₆O₂
MW: 156.22 g/mol
MDL Number: MFCD00038339
CAS Number: 123-68-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 0.06 g/l (20 °C)
Molar Mass 156.23 g/mol
Boiling Point 186 - 188 °C
Flash Point 74 °C
Density 0.887 g/cm3 (25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR