2-Amino-2-cyanoacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14902.0005 8.14902.0050 8.14902.0050 8.14902.0005
EM8.14902.0005CS 93123 USD
EM8.14902.0005 CA8.14902.0005 EM8.14902.0050 CA8.14902.0050
2-Amino-2-cyanoacetamide for synthesis, Sigma-Aldrich®
2-Amino-2-cyanoacetamide

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 119 - 122 °C
Molar Mass 99.09 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR