4-Aminohippuric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: p-Aminohippuric acid2-(4-Aminobenzamido)acetic acid

M-Clarity™ quality level = MQ200

8.14621.0025 8.14621.0025
EM8.14621.0025CS 20860 USD
EM8.14621.0025 CA8.14621.0025
4-Aminohippuric acid for synthesis, Sigma-Aldrich®
4-Aminohippuric acid
Formula: H₂NC₆H₄C(O)NHCH₂COOH
MW: 194.19 g/mol
MDL Number: MFCD00007890
CAS Number: 61-78-9

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 199 - 202 °C (decomposition)
Molar Mass 194.18 g/mol
Bulk Density 380 kg/m3
pH 3.0 - 3.5 (20 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR