(1S)-(+)-Camphorlactone sulfonyloxaziridine for synthesis, Sigma-Aldrich®

Synonyms: (1S,6R,9S)-12,12-DIMETHYL-5,8-DIOXA-3-THIA-4-AZATETRACYCLO[7.2.1.01,6.04,6]DODECAN-7-ONE 3,3-DIOXIDE5H-4a,7-Methano-4H,9H-oxazirino[2,3-b]oxepino[3,4-c]isothiazol-9-one, dihydro-10,10-dimethyl-, 3,3-dioxide, (4aS,7S,9aR)-

M-Clarity™ quality level = MQ200

8.14564.0001 8.14564.0001
EM8.14564.0001CS 155164.17 USD
CA8.14564.0001 EM8.14564.0001
(1S)-(+)-Camphorlactone sulfonyloxaziridine for synthesis, Sigma-Aldrich®
(1S)-(+)-Camphorlactone sulfonyloxaziridine
Formula: C₁₀H₁₃NO₅S
MW: 259.27 g/mol
MDL Number: MFCD00238755
CAS Number: 127411-75-0

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 202 - 205 °C
Molar Mass 259.27 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR