5-tert-Butyl-m-xylene for synthesis, Sigma-Aldrich®
Synonyms:
1-tert-Butyl-3,5-dimethylbenzene
M-Clarity™ quality level = MQ200
8.14287.0500
8.14287.0100
8.14287.0500
8.14287.0100
EM8.14287.0100CS
862.37
USD
CA8.14287.0500
EM8.14287.0100
EM8.14287.0500
CA8.14287.0100
5-tert-Butyl-m-xylene for synthesis, Sigma-Aldrich®
5-tert-Butyl-m-xylene
Formula:
C₁₂H₁₈ MW: 162.27 g/mol |
MDL Number:
MFCD00008832 CAS Number: 98-19-1 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | -22 °C |
Molar Mass | 162.27 g/mol |
Boiling Point | 207 - 209 °C (1013 hPa) |
Vapor Pressure | 0.4 hPa (20 °C) |
Flash Point | 84 °C DIN 51758 |
Density | 0.86 g/cm3 (20 °C) |
pH | 7 (H2O, 20 °C) |
Ignition Point | 560 °C DIN 51794 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...