Trimethylsilylacetylene for synthesis

Supplier: MilliporeSigma

Synonyms: Ethynyltrimethylsilane

M-Clarity™ quality level = MQ200

8.14226.0005 8.14226.0025 8.14226.0005 8.14226.0025
EM8.14226.0005CS 5149.18 USD
CA8.14226.0005 CA8.14226.0025 EM8.14226.0005 EM8.14226.0025
Trimethylsilylacetylene for synthesis
Trimethylsilylacetylene
Formula: C₅H₁₀Si
MDL Number: MFCD00008569
CAS Number: 1066-54-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 98.21 g/mol
Boiling Point 52 - 53 °C
Flash Point -34 °C
Refractive Index 1.3892 (20 °C, 589 nm)
Density 0.71 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR