Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

4-Aminobenzonitrile for synthesis

Synonyms: p-Cyanoaniline4-Cyanoanilinep-Aminobenzonitrile

8.14183.0010 8.14183.0010
EM8.14183.0010CS 29259.99 USD InStock
CA8.14183.0010 EM8.14183.0010
4-Aminobenzonitrile for synthesis
4-Aminobenzonitrile


Formula: C₇H₆N₂
MW: 118,14 g/mol
Boiling Pt: 167 °C (1.1 torr)
Melting Pt: 84 to 87 °C
Density: 1.274 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00007821
CAS Number: 873-74-5
EINECS: 212-850-1
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Melting Point 84 - 87 °C
Molar Mass 118.13 g/mol
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty