Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Terephthalaldehyde for synthesis

Supplier: MilliporeSigma

Synonyms: Terephthaldicarboxaldehydep-PhthalaldehydeTerephthaldialdehyde

8.08617.0100 8.08617.0100
EM8.08617.0100CS 1055.69 USD InStock
EM8.08617.0100 CA8.08617.0100
Terephthalaldehyde for synthesis
Terephthalaldehyde


Formula: C₈H₆O₂
MW: 134,13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Density: 1.381 g/cm³ (20 °C)
MDL Number: MFCD00006949
CAS Number: 623-27-8
EINECS: 210-784-8
Order Now
Specification Test Results
Solubility 3 g/l (50 °C)
Melting Point 112 - 115 °C
Molar Mass 134.13 g/mol
Boiling Point 245 - 248 °C (1013 hPa)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty