DL-Tropic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
2-Phenylhydracrylic acid
, (±)-Tropic acid
, 3-Hydroxy-2-phenylpropionic acid
, Tropic acid
, 3-Hydroxy-2-phenylpropanoic acid
M-Clarity™ quality level = MQ200
8.08384.0025
8.08384.0025
EM8.08384.0025CS
32073
USD
CA8.08384.0025
EM8.08384.0025
DL-Tropic acid for synthesis, Sigma-Aldrich®
DL-Tropic acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol |
MDL Number:
MFCD00004255 CAS Number: 529-64-6 |
Specification Test Results
Solubility | 20 g/l (20 °C) |
Melting Point | 115 - 118 °C |
Molar Mass | 166.17 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...