(S)-(+)-Mandelic acid for synthesis, for resolution of racemates, Sigma-Aldrich®

Danger

Synonyms: (S)-(+)-2-Hydroxy-2-phenylacetic acidL(+)-2-Hydroxy-2-phenylacetic acid(S)-2-Hydroxy-2-phenylacetic acidL(+)-Mandelic acid

M-Clarity™ quality level = MQ200

8.06925.0025 8.06925.0025 8.06925.0005 8.06925.0005
EM8.06925.0005CS 37935.31 USD
CA8.06925.0005 EM8.06925.0005 EM8.06925.0025 CA8.06925.0025
(S)-(+)-Mandelic acid for synthesis, for resolution of racemates, Sigma-Aldrich®
(S)-(+)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 130…132 °C
Density: 1.349 g/cm³ (20 °C)
Flash Pt: >190 °C
MDL Number: MFCD00004495
CAS Number: 17199-29-0
EINECS: 241-240-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 130 - 132 °C
Molar Mass 152.15 g/mol
Bulk Density 670 kg/m3
Flash Point 190 °C
pH 2.1 (60 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR