Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Nitronaphthalene for synthesis

Supplier: MilliporeSigma

Synonyms: α-Nitronaphthalene

8.06291.0500 8.06291.0100 8.06291.0100 8.06291.0500
EM8.06291.0500CS 351.08 USD InStock
EM8.06291.0500 EM8.06291.0100 CA8.06291.0100 CA8.06291.0500
1-Nitronaphthalene for synthesis
1-Nitronaphthalene


Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 59 - 61 °C
Density: 1.331 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 86-57-7
EINECS: 201-684-5
UN: 2538
ADR: 4.1,III
Merck Index: 12,06711
Order Now
Specification Test Results
Solubility 0.022 g/l (20 °C)
Melting Point 59 - 61 °C
Molar Mass 173.17 g/mol
Boiling Point 304 °C
Vapor Pressure 0.00015 hPa (20 °C)
Flash Point 170 °C DIN 51758
Density 1.33 g/cm3 (20 °C)
Ignition Point 450 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty