Dibenzosuberone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
5-Dibenzosuberone
, 1-Dibenzo[b,f]cycloheptanone
, Dibenzo[b,f]cycloheptan-1-one
, 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one
M-Clarity™ quality level = MQ200
8.03559.0050
8.03559.0050
EM8.03559.0050CS
9422
USD
CA8.03559.0050
EM8.03559.0050
Dibenzosuberone for synthesis, Sigma-Aldrich®
Dibenzosuberone
Formula:
C₁₅H₁₂O MW: 208.26 g/mol Boiling Pt: 378 °C (1013 hPa) Melting Pt: 31…34 °C Density: 1.1635 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003587 CAS Number: 1210-35-1 EINECS: 214-912-3 |
Specification Test Results
Solubility | 0.03 g/l (20 °C) |
Melting Point | 31 - 34 °C |
Molar Mass | 208.25 g/mol |
Boiling Point | 191 - 194 °C (7 hPa) |
Flash Point | 200 °C |
Refractive Index | 1.6324 (20 °C, 589 nm) |
Density | 1.16 g/cm3 (25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...