3-Bromotoluene for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.02400.0010 8.02400.0050 8.02400.0050 8.02400.0010
EM8.02400.0050CS 11816.25 USD
EM8.02400.0050 CA8.02400.0010 CA8.02400.0050 EM8.02400.0010
3-Bromotoluene for synthesis, Sigma-Aldrich®
3-Bromotoluene
Formula: C₇H₇Br
MW: 171.04 g/mol
Boiling Pt: 183.7 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.41 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000085
CAS Number: 591-17-3
EINECS: 209-702-3
UN: 1993
ADR: 3,III
Merck Index: 13,01424

Order Now

Specification Test Results

Solubility 0.05 g/l
Melting Point -40 °C
Molar Mass 171.03 g/mol
Boiling Point 184 °C
Vapor Pressure 2.04 hPa (25 °C)
Flash Point 60 °C
Refractive Index 1.5517 (20 °C, 589 nm)
Density 1.405 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR