Benzamide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.02191.0005
8.02191.0250
8.02191.0250
EM8.02191.0250CS
2403
USD
EM8.02191.0250
CA8.02191.0250
EM8.02191.0005
Benzamide for synthesis, Sigma-Aldrich®
Benzamide
Formula:
C₇H₇NO MW: 121.14 g/mol Boiling Pt: 288 °C (1013 hPa) Melting Pt: 124 °C Density: 1.35 g/cm³ (20 °C) Flash Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007968 CAS Number: 55-21-0 EINECS: 200-227-7 Merck Index: 13,01059 |
Specification Test Results
Solubility | 13 g/l (25 °C) |
Melting Point | 124 - 127 °C |
Molar Mass | 121.14 g/mol |
Bulk Density | 550 kg/m3 |
Vapor Pressure | 0.000056 hPa (20 °C) |
Flash Point | 180 °C |
Density | 1.34 g/cm3 (20 °C) |
pH | 6.9 (H2O) (saturated solution) |
Ignition Point |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...