Dicyclohexyl phthala for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Dicyclohexyl phthalate

M-Clarity™ quality level = MQ200

8.00920.0100 8.00920.1000 8.00920.0100 8.00920.1000
EM8.00920.1000CS 679 USD
EM8.00920.1000 EM8.00920.0100 CA8.00920.1000 CA8.00920.0100
Dicyclohexyl phthala for synthesis, Sigma-Aldrich®
Dicyclohexyl phthala
Formula: C₂₀H₂₆O₄
MW: 330.42 g/mol
MDL Number: MFCD00003849
CAS Number: 84-61-7
EINECS: 201-545-9

Order Now

Specification Test Results

Solubility 0.2 mg/l (20 °C)
Melting Point 62 - 64 °C
Molar Mass 330.42 g/mol
Bulk Density 710 kg/m3
Boiling Point 226 °C (1013 hPa)
Vapor Pressure 0.0000039 hPa (50 °C)
Flash Point 180 - 190 °C
Density 1.15 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 395 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR