Picolinic acid for synthesis, Sigma-Aldrich®

Warning

Synonyms: 2-Pyridinecarboxylic acid2-Picolinic acidPyridine-2-carboxylic acid

M-Clarity™ quality level = MQ200

8.00735.0100 8.00735.0100
EM8.00735.0100CS 1070.26 USD
CA8.00735.0100 EM8.00735.0100
Picolinic acid for synthesis, Sigma-Aldrich®
Picolinic acid
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 134…137 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557

Order Now

Specification Test Results

Solubility 887 g/l (20 °C)
Melting Point 134 - 137 °C
Molar Mass 123.11 g/mol
Bulk Density 500 - 700 kg/m3
Flash Point 139 °C
pH 3.1 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR