Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aminoacetaldehyde dimethyl acetal for synthesis


8.00368.0100 8.00368.0100 8.00368.0025 8.00368.0025
EM8.00368.0100CS 2268.41 USD InStock
CA8.00368.0100 EM8.00368.0100 CA8.00368.0025 EM8.00368.0025
Aminoacetaldehyde dimethyl acetal for synthesis
Aminoacetaldehyde dimethyl acetal


Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
MDL Number: MFCD00008135
CAS Number: 22483-09-6
EINECS: 245-026-5
Order Now
Specification Test Results
Solubility (20 °C) soluble
Molar Mass 105.13 g/mol
Boiling Point 135 - 139 °C (126 hPa)
Flash Point 41 °C
Refractive Index 1.417 (20 °C, 589 nm)
Density 0.97 g/cm3 (20 °C)
pH (H2O) (undiluted),alkaline

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty