Indium ≥99%, granular, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.12196.0100 1.12196.0100
EM1.12196.0100CS 147258 USD
EM1.12196.0100 CA1.12196.0100
Indium ≥99%, granular, Sigma-Aldrich®
Indium
Formula: In
MW: 114.82 g/mol
Density: 7.31 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
ADR: 4.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 156 °C
Molar Mass 114.82 g/mol
Boiling Point 2000 °C
Density 7.31 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR