Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Trichloroethylene, EMSURE® ACS, Reag. Ph. Eur. for analysis

Supplier: MilliporeSigma

Danger
Synonyms: 1,1,2-Trichloroethene1,1,2-TrichloroethyleneTrichloroethene

1.11872.2500 1.11872.1000 1.11872.1000 1.11872.2500
EM1.11872.1000CS 1235.81 USD InStock
CA1.11872.2500 CA1.11872.1000 EM1.11872.1000 EM1.11872.2500
Trichloroethylene, EMSURE® ACS, Reag. Ph. Eur. for analysis
Trichloroethylene


Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 86.7 °C (1013 hPa)
Melting Pt: -73 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Merck Index: 12,09769
https://us.vwr.com/stibo/web/std.lang.all/13/81/741381.jpg
Order Now
Specification Test Results
Solubility 1.28 g/l (25 °C)
Melting Point -86 °C
Molar Mass 131.39 g/mol
Vapor Pressure 77 hPa (20 °C)
Explosion Limit
Density 1.46 g/cm3 (20 °C)
Ignition Point 410 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty