Tetrabutylammonium hydroxide solution 0.1 mol/L (0.1 N), Titripur®, Supelco®

Danger

1.09162.0500
CA1.09162.0500
Tetrabutylammonium hydroxide solution 0.1 mol/L (0.1 N), Titripur®, Supelco®
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Flash Point 12 °C
Density 0.792 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR