Phosphinic acid (Hypophosphorous acid) 50%, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.04633.0500
EM1.04633.0500CS 3535 USD
EM1.04633.0500
Phosphinic acid (Hypophosphorous acid) 50%, EMSURE® for analysis, Supelco®
Phosphinic acid (Hypophosphorous acid)
Formula: H₃O₂P
MDL Number: MFCD02183592
CAS Number: 6303-21-5
EINECS: 228-601-5
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 108 °C (1013 hPa) (decomposition)
Vapor Pressure 30 hPa (20 °C)
Density 1.21 - 1.26 g/cm3 (20 °C)
pH 1 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR