Mercury(II) sulfate ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Mercury sulfateMercuric sulphateMercury sulphateMercuric sulfate

1.04480.0250 1.04480.0050 1.04480.0250 1.04480.0050
EM1.04480.0050CS 23907 USD
EM1.04480.0050 CA1.04480.0250 CA1.04480.0050 EM1.04480.0250
Mercury(II) sulfate ACS for analysis, Supelco®
Mercury (II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850 °C
Density: 6.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1,II
Merck Index: 13,05914

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 296.65 g/mol
Bulk Density 670 kg/m3
Density 6.47 g/cm3 (20 °C)
pH 1 (50 g/l, H2O, 20 °C)
Ignition Point 450 °C
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR