Mercury(II) nitrate monohydrate ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Mercury(II) nitrate hydrate
, Mercury dinitrate monohydrate
M-Clarity™ quality level = MQ300
1.04439.0050
1.04439.0050
1.04439.0250
1.04439.0250
EM1.04439.0250CS
39114
USD
CA1.04439.0250
EM1.04439.0250
EM1.04439.0050
CA1.04439.0050
Mercury(II) nitrate monohydrate ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) nitrate monohydrate
Formula:
Hg(NO₃)₂·1H₂O MW: 342.62 g/mol Melting Pt: 79 °C Density: 4.3 g/cm³ (20 °C) |
MDL Number:
MFCD00149736 CAS Number: 7783-34-8 EINECS: 233-152-3 UN: 1625 ADR: 6.1,II |
Specification Test Results
Assay (complexometric) | ≥ 99.0 % |
Chloride (Cl) | ≤ 0.002 % |
Sulfate (SO₄) | ≤ 0.002 % |
Cu (Copper) | ≤ 0.0005 % |
Fe (Iron) | ≤ 0.001 % |
K (Potassium) | ≤ 0.005 % |
Na (Sodium) | ≤ 0.005 % |
Residue after reduction | ≤ 0.01 % |
Corresponds to ACS,Reag. Ph Eur |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...