Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Glycerine ≥85%, EMSURE® Reag. Ph. Eur. for analysis

Supplier: MilliporeSigma

Synonyms: GlycerinGlysaninGlyceritolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

1.04094.1000 1.04094.9026 1.04094.9026 1.04094.0500 1.04094.1000 1.04094.2500 1.04094.2500 1.04094.0500
EM1.04094.9026EA 945.5 USD InStock
CA1.04094.1000 EM1.04094.9026 CA1.04094.9026 CA1.04094.0500 EM1.04094.1000 EM1.04094.2500 CA1.04094.2500 EM1.04094.0500
Glycerine ≥85%, EMSURE® Reag. Ph. Eur. for analysis
Glycerine


Formula: HOCH₂CH(OH)CH₂OH
MW: 92,09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C ()
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
UN: 0000
Merck Index: 13,04497
https://us.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -10 °C
Boiling Point 130 °C (1013 hPa)
Vapor Pressure 8 hPa (20 °C)
Flash Point 199 °C
Density 1.23 g/cm3 (20 °C)
pH 5 (100 g/l, H2O, 20 °C)
Ignition Point 429 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty