Glycerine, EMSURE® ACS, Reag. Ph. Eur. for analysis

Supplier: MilliporeSigma

Synonyms: GlysaninGlycerinGlyceritolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

1.04092.4000 1.04092.9010 1.04092.4000 1.04092.9010 1.04092.1000 1.04092.1000 1.04092.2500
EM1.04092.4000EA 800.02 USD InStock
EM1.04092.4000 EM1.04092.9010 CA1.04092.4000 CA1.04092.9010 CA1.04092.1000 EM1.04092.1000 CA1.04092.2500
Glycerine, EMSURE® ACS, Reag. Ph. Eur. for analysis
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.1 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18,6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C ()
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
UN: 0000
Merck Index: 13,04497
https://us.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 18 °C
Molar Mass 92.1 g/mol
Boiling Point 290 °C (1013 hPa) (decomposition)
Vapor Pressure  - 0.001 hPa (20 °C)
Flash Point 199 °C
Explosion Limit 2.6 - 11.3 %(V)
Density 1.26 g/cm3 (20 °C)
pH 5 (100 g/l, H2O, 20 °C)
Ignition Point 400 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR