Celite® 503, filter aid, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02692.1000
1.02692.1000
89501-600EA
304.36
USD
CA89501-600
89501-600
Celite® 503, filter aid, Supelco®
Celite® 503, filter aid
Formula:
O₂Si MW: 60.08 g/mol |
CAS Number:
68855-54-9 |
Specification Test Results
Solubility |
|
Bulk Density | 300 kg/m3 |
pH | 10 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...