Celite® Hyflo Super-cel®, filter aid, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02688.1000
1.02688.1000
89501-608EA
193.62
USD
89501-608
CA89501-608
Celite® Hyflo Super-cel®, filter aid, Supelco®
Celite® Hyflo® Super Cel, filter aid
Boiling Pt:
2200 °C (1013 hPa) Melting Pt: 1710 °C Density: 2.3 g/cm³ (20 °C) |
MDL Number:
MFCD00132803 CAS Number: 68855-54-9 |
Specification Test Results
Solubility |
|
Bulk Density | 300 kg/m3 |
pH | 10 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...