Lead borate, Supelco®

Supplier: MilliporeSigma
Danger

1.01189.1000 1.01189.1000
EM1.01189.1000CS 72194.99 USD
EM1.01189.1000 CA1.01189.1000
Lead borate, Supelco®
Lead borate
Formula: B₂O₆Pb₃
CAS Number: 35498-15-8
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (25 °C) soluble
Molar Mass 328.85 g/mol
Bulk Density 3870 kg/m3
Density 11.0 g/cm3 (20 °C)
pH 8.6 (650 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR