Sulfuric acid 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.00732.2500
1.00732.2500
EM1.00732.2500CS
1175
USD
EM1.00732.2500
CA1.00732.2500
Sulfuric acid 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 g/mol Boiling Pt: ∼335 °C (1013 hPa) Melting Pt: 3 °C Density: 1.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II Merck Index: 14,08974 |
Specification Test Results
Ag (Silver) | ≤ 0.02 ppm |
Al (Aluminium) | ≤ 0.05 ppm |
Appearance | passes test |
As (Arsenic) | ≤ 0.01 ppm |
Assay (alkalimetric) | 95.0 - 97.0 % |
Ba (Barium) | ≤ 0.05 ppm |
Be (Beryllium) | ≤ 0.01 ppm |
Bi (Bismuth) | ≤ 0.1 |
Ca (Calcium) | ≤ 0.2 ppm |
Cd (Cadmium) | ≤ 0.02 ppm |
Chloride (Cl) | ≤ 0.1 ppm |
Co (Cobalt) | ≤ 0.01 ppm |
Color | ≤ 10 Hazen |
Cr (Chromium) | ≤ 0.05 ppm |
Cu (Copper) | ≤ 0.01 ppm |
Date of expiry | see product label |
Density (d 20/20) | 1.834 - 1.837 |
Fe (Iron) | ≤ 0.1 |
Ge (Germanium) | ≤ 0.05 ppm |
Heavy metals (as Pb) | ≤ 1 ppm |
Hg (Mercury) | ≤ 0.005 ppm |
Identity | passes test |
K (Potassium) | ≤ 0.1 |
Li (Lithium) | ≤ 0.01 ppm |
Mg (Magnesium) | ≤ 0.05 ppm |
Mn (Manganese) | ≤ 0.01 ppm |
Mo (Molybdenum) | ≤ 0.05 ppm |
Na (Sodium) | ≤ 0.5 ppm |
NH₄ (Ammonium) | ≤ 2 ppm |
Ni (Nickel) | ≤ 0.02 ppm |
Nitrate (NO₃) | ≤ 0.2 ppm |
Pb (Lead) | ≤ 0.02 ppm |
Phosphate (PO₄) | ≤ 0.5 ppm |
Residue on ignition | ≤ 5 ppm |
Sr (Strontium) | ≤ 0.02 ppm |
Substances reducing potassium permanganate (as SO₂) | ≤ 2 ppm |
Ti (Titanium) | ≤ 0.1 |
Tl (Thallium) | ≤ 0.05 ppm |
V (Vanadium) | ≤ 0.01 ppm |
Zn (Zinc) | ≤ 0.05 ppm |
Zr (Zirconium) | ≤ 0.1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...