Hydriodic acid 57%, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hydrogen iodide
, Hydroiodic acid
M-Clarity™ quality level = MQ300
1.00344.0250
1.00344.1000
EM1.00344.0250CS
7054.02
USD
EM1.00344.0250
EM1.00344.1000
Hydriodic acid 57%, EMSURE® for analysis, Supelco®
Hydriodic acid
Formula:
HI MW: 127.91 g/mol Boiling Pt: 127 °C (1013 hPa) Density: 1.7 g/cm³ (20 °C) |
MDL Number:
MFCD00011347 CAS Number: 10034-85-2 EINECS: 233-109-9 UN: 1787 ADR: 8,II |
Specification Test Results
Al (Aluminium) | ≤ 1 ppm |
Alkoxygroups | passes test |
As (Arsenic) | ≤ 1 ppm |
Assay (acidimetric) | ≥ 57 % |
Ba (Barium) | ≤ 0.1 ppm |
Be (Beryllium) | ≤ 0.1 ppm |
Bi (Bismuth) | ≤ 0.1 ppm |
Ca (Calcium) | ≤ 1 ppm |
Cd (Cadmium) | ≤ 0.1 ppm |
Chloride and Bromide (as Cl) | ≤ 100 ppm |
Co (Cobalt) | ≤ 0.1 ppm |
Cr (Chromium) | ≤ 0.1 ppm |
Cu (Copper) | ≤ 0.1 ppm |
Fe (Iron) | ≤ 2 ppm |
Ge (Germanium) | ≤ 0.1 ppm |
K (Potassium) | ≤ 1 ppm |
Li (Lithium) | ≤ 0.1 ppm |
Mg (Magnesium) | ≤ 5 ppm |
Mn (Manganese) | ≤ 0.1 ppm |
Mo (Molybdenum) | ≤ 0.1 ppm |
Na (Sodium) | ≤ 1 ppm |
Ni (Nickel) | ≤ 0.1 ppm |
Pb (Lead) | ≤ 0.1 ppm |
Phosphorous compounds (as PO₄) | ≤ 10 ppm |
Residue on ignition (as sulfate) | ≤ 50 ppm |
Sr (Strontium) | ≤ 0.1 ppm |
Sulfate (SO₄) | ≤ 20 ppm |
Ti (Titanium) | ≤ 0.1 ppm |
Tl (Thallium) | ≤ 0.1 ppm |
V (Vanadium) | ≤ 0.1 ppm |
Zn (Zinc) | ≤ 0.5 ppm |
Zr (Zirconium) | ≤ 0.1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...