Hydriodic acid 57%, Extra Pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hydrogen iodide
, Hydroiodic acid
M-Clarity™ quality level = MQ200
1.00341.0250
1.00341.9022
EM1.00341.9022EA
19842
USD
EM1.00341.9022
EM1.00341.0250
Hydriodic acid 57%, Extra Pure, Supelco®
Hydriodic acid
Formula:
HI MW: 127.91 g/mol Boiling Pt: 127 °C (1013 hPa) Density: 1.7 g/cm³ (20 °C) |
MDL Number:
MFCD00011347 CAS Number: 10034-85-2 EINECS: 233-109-9 UN: 1787 ADR: 8,II |
Specification Test Results
Assay (acidimetric) | ≥ 57 % |
Chloride and Bromide (as Cl) | ≤ 0.03 % |
Fe (Iron) | ≤ 0.0005 % |
Heavy metals (as Pb) | ≤ 0.001 % |
Non volatile matter | ≤ 0.06 % |
Sulfate (SO₄) | ≤ 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...