Would you like to log in again?
Show Me Science Chemistry & Physics - Physics: Gravity and Forces
Supplier: TMW MEDIA GROUP SE
K4418DD K4418DVD
The 9-title Wonders of Physics and Chemistry series covers everything from the definition of an atom to the science of light. Students can start to develop basic understandings of the fundamentals of science, then work their way up to more complex subjects. Each program includes a subject specific teachers guide which includes subject matter revision and suggestions on how to present material, questions to ask students, and class projects to help illustrate the key concepts. One of Newton’s Laws of Motion states that if the forces acting on an object are balanced, then the object will maintain a constant speed of travel. For example, if friction and air resistance acting upon a cyclist are equal to the pedaling speed, the cyclist’s speed will remain the same. This program explains forces such as lift, air resistance, and gravity, and their effects on skydivers, airplanes, and racing cars. The program also describes how airplanes counter gravity in order to fly.
**Please note this is a Digital Product**
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Gravity And Forces DVD Version K4418DVD 470123-238 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty