SCIENCEWARE® Poxygrid® Eye Wash Bottle Wall Rack, Bel-Art
Supplier: Bel-Art Products
Poxygrid®
H24859-0000
56611-083EA 66.05 USD InStock
56611-083
SCIENCEWARE® Poxygrid® Eye Wash Bottle Wall Rack, Bel-Art
Racks Bottle Racks
Fabricated of welded steel wire with corrosion-proof epoxy coating, this wall rack holds a single 1000 mL (32oz.) eye wash bottle in plain view for emergency use.
  • Welded steel wire construction
  • Corrosion-proof epoxy coating
  • Suitable for use with sterile eye wash
  • Kept in plain view for emergency use
It hangs on a screw or nail, and is suitable for use with sterile saline eye wash (56611-043) and eye wash bottles (56611-075). Eye wash and bottles are sold separately.