Biotix Plate Sealing Mats, Biotix
MS-DWP-9SP
89511-238CS 360.22 USD
89511-238
Biotix Plate Sealing Mats, Biotix
Specially designed for Biotix deep well and assay plates, these reusable sealing mats are cost effective and ideal for sample processing and storage applications
Biotix sealing mats are made of high quality research-grade silicone, giving them the ability to withstand repeated autoclaving, bleach, and ethanol rinses. Mats for 96-Well plates are pre-scored for piercing with standard pipet tips. The 384-Well style sealing mats are piercable with a syringe. All mats can support use in a wide temperature range, from –80 to 121°C.
Ordering information: All sealing mats are packaged 10 mats per pack, 5 packs per case.