Would you like to log in again?
Biotix PCR Tubes, Biotix
Supplier: Biotix
PCT-0002-1CT PCT-0002-8C PCT-0002-8CF
Biotix PCR tubes are manufactured under stringent QC protocols with medical-grade virgin polypropylene.
  • Precision engineered caps which prevent leaking and evaporative effects when heated
  • Ideal for qPCR with the 8 strip format
  • Certified free of DNase, RNase, endotoxins (pyrogens) and PCR inhibitors
  • Flat cap which is easy to open and close one handed
  • These tubes click closed so you know you’ve made a secure seal
Biotix PCR tubes are manufactured under stringent QC protocols with medical-grade virgin polypropylene. These tubes consist of a thin and uniform wall thickness that provides equal thermal transfer though the tube for optimal amplification.
ORDER
Expand All / Collapse All
Description Volume Color Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Individual PCR Tube, Thin Wall, Flat Cap 0.2 ml Clear 1,000 Tubes/Pack, 10 Packs/Case PCT-0002-1CT 89511-270 Case of 10000 Retrieving
Pack of 1,000 Retrieving
8-Strip PCR Tubes 0.2 mL Clear 125 Tubes/Pack, 10 Packs/Case PCT-0002-8C 89511-272 Case of 1250 Retrieving
Pack of 125 Retrieving
8-Strip Flat Caps for PCR Clear 125 Caps/Pack, 10 Packs/Case PCT-0002-8CF 89511-274 Case of 1250 Retrieving
Pack of 125 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty