Biotix PCR Plates, Biotix
P-0200-9CFS
89511-280CS 414.66 USD
89511-280
Biotix PCR Plates, Biotix
Developed for use with the industry's most popular thermocyclers, Biotix PCR plates are manufactured and tested to ensure robust and reproducible amplification under the most exacting protocols.
Each plate offers a molded-in alphanumeric grid to help with sample identification, and can withstand centrifugation up to 4,000 RCF when supported by the appropriate plate rotor adaptor. Slightly raised well rims facilitate secure sealing with films or strip caps.
As with all Biotix products, PCR plates are free from heavy metals, mold releases, or other extractables that could interfere with highly sensitive protocols. They are BioReady™ Certified for purity.
Made from medical-grade virgin polypropylene, these plates feature ultra-thin wall wells that provide excellent thermal transfer with PCR heat blocks.
Certifications: CE certified. IVD labeled. Certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.