Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Triumph Board Projector PJ3000 UST-WDLP
8592580081989
470019-144BX 1729.25 USD InStock
470019-144
Triumph Board Projector PJ3000 UST-WDLP
Furniture Classroom Furniture
This wide-screen, ultra short throw projector eliminates shadow and glare due to close proximity of projector to board. Offering 2D and 3D projection. Specifications: Native Resolution WXGA 1280 x 800; Brightness 3500 ANSI Lumens, Contrast 15000:1; Noise Level 30dB (ECO mode); Aspect Ratio 16:10 Native, 16:9/4:3 Compatible; Weight <3.0kg, Inputs/Outputs: VGA in x 2, VGA out x 1, HDMI x 1, S-Video x 1, Composite x 1, Audio in –RCA (L/R) x 2, Audio in – 3.5mm jack x 2, Audio out x 1, microphone x 1, Mini USB x 1, DC 12V output x 1, Control RS232 D-Sub 9pin x 1, RJ45 (optional), Speakers 10W.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty