Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Triumph Board MULTI Touch
Supplier: TRIUMPH BOARD A.S.
8592580101458 8592580101472 8592580101465
470214-256BX 1173.69 USD InStock
470214-256 470214-260 470214-258
Triumph Board MULTI Touch
Furniture Classroom Furniture
This interactive whiteboard allows four users to write, draw, move objects or two users to resize and rotate two objects with two fingers per the object simultaneously. 
  • Based on IR Cell LED Optical Technology
  • Multi-Touch Unit
  • Multi-User Mode Setting
It is a truly multi-touch, requiring no separate multi-user mode setting. Users can write, draw and move objects with a finger, stylus, or even a pointer. It connects to a computer through a fast USB 2.0 interface or optionally with wireless radio technology with 2. 4GHz. It supports Linux, Mac, all Microsoft Windows and multi-user writing and common multi-touch gestures currently recognized in Microsoft Win-7. Use simple intuitive finger or hand gestures to toss, rotate, or zoom in and out on objects.
The unit is based on IR Cell LED Optical technology, which uses a combination of IR LEDs and photo transistors that are aligned in pairs around the active surface. These pairs of LEDs and photo transistors cast a grid over this surface. When the surface is touched, the beam of infrared light between the LEDs and the photo transistors is interrupted at the point of touch. This position is converted into an ‘X’ and a ‘Y’ coordinate which the computer then converts into a mouse position.
Ordering information: Purchase price does not include installation. To learn more about installation opportunities, please contact your VWR Sales Representative.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty