Would you like to log in again?
Freeze/Thaw Chambers, Caron Products
89186-108EA 22874.95 USD InStock
89186-108 89186-096 89186-098 89186-106 89186-110 89186-104 89186-100 89186-102
Freeze/Thaw Chambers, Caron Products
Testing Chambers Temperature Test Chambers
Caron’s Freeze/Thaw Chambers precisely control a wide range of temperature and (optional) humidity settings
Chambers will accommodate a wide range of freeze/thaw studies, including pharmaceutical drug studies, concrete and asphalt testing, coating and adhesive testing, outdoor weathering tests, stress testing, and more. The chambers include a deluxe controller that will ramp and soak to cycle freezing and thawing for repeatable, accurate studies. The controller can perform 4 programs, 40 individual steps, an infinite number of loops, and a ramp based on any time or rate. The deluxe controller comes standard with analog outputs and RS-485 communications. It connects the unit directly to an in-house monitoring system. Caron offers an optional factory calibration of the controller and will program it to specific user profiles, which makes installation and start-up easy.
This product offering includes two sizes of large capacity, reach-in units. The chambers are not only consistently reliable, but offer best-in-class customer friendly features and unique energy efficient components. Multiple optional accessories are available, allowing users to customize the chamber to specific needs.
Models 6220-2, 6220-3, 6240-2, and 6240-3 offer a relative humidity range of 20 to 98%, with ±3% relative humidity control. These units are equipped with a capacitive humidity sensor. All shelving is constructed of electropolished Type 304 Perforated Stainless Steel.
This broad range encompasses nearly every ambient environment on earth. Automatic, versatile, and programmable cycling between setpoints enables continuous simulation of the harsh conditions of freezing and thawing for rapid testing. These demanding temperature and humidity cycling studies require accurate control of thermal conditions, and are best served by our chambers.
Certifications: All 230V units are CE marked.
Ordering information: The 708L (25cu.ft.) units are supplied complete with four shelves. The 934L (33cu.ft.) units are supplied complete with five shelves.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty