ISOTON® II Diluent, Beckman Coulter

Supplier: Beckman Coulter
8546719 8546719
BK8546719EA 107.85 USD
BK8546719 CABK8546719
ISOTON® II Diluent, Beckman Coulter
Buffers Biological Buffers
Beckman Coulter™ ISOTON™ II Diluent is a filtered, phosphate-buffered saline solution compatible with human blood cells, and may be used for the suspension of most biological cells and many industrial samples.

  • Dilution Agent
  • For use with Flow Cytometers
  • pH 7.35-7.65

ISOTON™ II Diluent is a necessary reagent for the daily maintenance and operation of Beckman Coulter flow cytometers, including Multisizer particle counter appliances. The Multisizer is a highly versatile particle counting and characterization system. It uses the Coulter principle to detect particles via electrical zone sensing, regardless of the nature of the particle or its optical properties. This makes it an ideal tool for detecting and counting a wide variety of particles.

This flow cytometry test kit is 1 L x 20 L.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR