Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Potato Dextrose Agar, HiMedia
Supplier: HiMedia
M096-500G MU096-500G M096-500G M096-5KG MU096-500G M096-5KG
95022-794EA 219.03 USD InStock
CA95020-250 95022-794 95020-250 95026-810 CA95022-794 CA95026-810
Potato Dextrose Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Cream to yellow homogeneous free flowing powder.
  • Recommended for the isolation and enumeration of yeasts and moulds
  • Free flowing powder
Potato dextrose agar is recommended for the isolation and enumeration of yeasts and moulds from dairy and other food products.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty