Water Molecular biology grade, sterile

Supplier: HiMedia

Synonyms: Dihydrogen oxideIce

TCL016-500ML TCL018-500ML TCL018-500ML TCL016-500ML TCL018-500ML
76203-540EA 14.32 USD InStock
CA95016-374 76203-540 CA95016-376 95016-374 95016-376
Water Molecular biology grade, sterile
Water

This water is molecular biology grade.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
UN: 0000
Merck Index: 13,10098

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR