Masterclave Automated Culture Media Preparator, bioMerieux
AESAP1081 AESAP1080 AESAP1067
97059-214EA 56676.64 USD InStock
97059-214 97059-212 97059-216
Masterclave Automated Culture Media Preparator, bioMerieux
Media Preparators
The Masterclave series Culture Media Preparators enable automated and quick preparation of the highest-quality culture media.
  • Automated, quick preparation
  • Excellent medium homogenization
  • Accurate core temperature monitoring
A large magnetic stirrer (patented) provides excellent medium homogenization during sterilization and distribution, and accurate core temperature monitoring guarantees highest-quality agar or broth even for large-prepared volumes.
Ordering information: These items require training and install by the vendor which is not included in the cost