Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Lactophenol blue solution stain (fungal)

Supplier: MilliporeSigma

Danger
Synonyms: Lactophenol anilin blue solutionLactophenol cotton blue solution

R03465-70 R03465-70
EM-R03465-70CS 1457.88 USD InStock
EM-R03465-70 CAR03465-70
Lactophenol blue solution stain (fungal)
Lactophenol blue solution
Lactophenol Cotton Blue is a mounting fluid for demonstrating the microscopic morphology of fungi.

  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use


Density: 1.16 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00146574
UN: 2821
ADR: 6.1,II
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty