Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
PBS, Phosphate Buffered Saline Tablets, Diagnostic Biosystem
Supplier: Diagnostic Biosystems
DMRE404-01
99990-026EA 149.61 USD InStock
CA99990-026 99990-026
PBS, Phosphate Buffered Saline Tablets, Diagnostic Biosystem
Buffers Electrophoresis Buffers Washing Buffers
White concave tablets
Dissolve one tablet in 100 mL of deionized water. Each tablet will make 100 mL of 10 mM (1x) PBS solution. The pH of the final solution will be 7.4 ± 0.1. For In Vitro Diagnostic Use. Store at room temperature. Protect from moisture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty