Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II) (13,3% Pd), crystalline solid

Warning

Synonyms: PdCl2(BINAP)

46588-06 46588-03 46588-06 46588-03
AA46588-06EA 648.24 USD
AA46588-06 CAAA46588-03 CAAA46588-06 AA46588-03
Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II) (13,3% Pd), crystalline solid
Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
Formula: C₄₄H₃₂Cl₂P₂Pd
MW: 800.01 g/mol
Melting Pt: 230…300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075254
CAS Number: 253157-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR